Anti-PAPD5 Goat Polyclonal Antibody

Supplier: ProSci
42-364
10113-824EA 701.94 USD
10113-824
Anti-PAPD5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 70 kDa band observed in mouse and rat spleen lysates (calculated MW of 69.4 kDa according to mouse NP_001157971.1. Recommended concentration: 0.2-0.6 ?g/mL.

PAPD5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PAPD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR