Anti-NCAM1 Goat Polyclonal Antibody

Supplier: ProSci
42-361
10114-384EA 701.94 USD
10114-384
Anti-NCAM1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 170 kDa band observed in human brain (cerebellum, cerebral and frontal cortex) lysates (calculated MW of 94.6 kDa according to NP_851996.2). The observed molecular weight corresponds to the glycosylated form. Recommended concentration: 0.01-0.03 ?g/mL.

NCAM1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NCAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR