Anti-C22ORF28 Goat Polyclonal Antibody

Supplier: ProSci

42-360
10113-822EA 560.36 USD
10113-822
Anti-C22ORF28 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 55 kDa band observed in HeLa, Jurkat and K562 lysate (calculated MW of 55.2 kDa according to NP_055121.1). Recommended concentration: 0.1-0.3 ?g/mL.

C22orf28 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: C22orf28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR