Anti-NUP84 Goat Polyclonal Antibody

Supplier: ProSci

42-354
10114-380EA 560.36 USD
10114-380
Anti-NUP84 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 75 kDa band observed in S. cerevisiae wildtype lysates but not in the knock-out strain (calculated MW of 83.6 kDa according to NP_010167.1). Recommended concentration: 2-4 ?g/mL.

NUP84 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NUP84
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR