Anti-TPH2 Goat Polyclonal Antibody

Supplier: ProSci
42-339
10113-796EA 701.94 USD
10113-796
Anti-TPH2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 85 kDa band in mouse fetal brain lysates after 0.3 ?g/mL antibody staining.9 kDa according to NP_775567.2. The 85 kDa band was successfully blocked by incubation with the immunizing peptide.

TpH 2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Tph2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR