Anti-MUC5AC Goat Polyclonal Antibody

Supplier: ProSci
42-329
10112-334EA 552.17 USD
10112-334
Anti-MUC5AC Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: This antibody stained cilia in mouse bronchi and it co-stained with lectin (membrane).

Muc5ac antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Muc5ac
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR