Anti-MUC19 Goat Polyclonal Antibody

Supplier: ProSci
42-309
10113-770EA 701.94 USD
10113-770
Anti-MUC19 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. Positive staining in the sublingual salivary gland of the mouse, while cells remain negative in the submandibular salivary gland.

Muc19 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Muc19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR