Anti-SGCG Goat Polyclonal Antibody

Supplier: ProSci

42-293
10114-330EA 455.3 USD
10114-330
Anti-SGCG Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 45 kDa band in human heart lysates after 1 ?g/mL antibody staining.4 kDa according to NP_000222.1. The 45 kDa band was successfully blocked by incubation with the immunizing peptide.

Gamma Sarcoglycan antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SGCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR