Anti-DIAPH1 Goat Polyclonal Antibody

Supplier: ProSci
42-285
10114-316EA 701.94 USD
10114-316
Anti-DIAPH1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 140 kDa band observed in lysates of cell line NIH3T3 (calculated MW of 141 kDa according to human NP_005210.3 and 139 kDa according to mouse NP_031884.1). Recommended concentration: 0.3-1 ?g/mL.

DIApH 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DIAPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR