Anti-CSMD1 Goat Polyclonal Antibody

Supplier: ProSci

42-276
10114-302EA 560.36 USD
10114-302
Anti-CSMD1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 150 kDa band observed in human cerebellum lysates (calculated MW of 389 kDa according to NP_150094.4). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Kraus et al, J Immunol. 2006 Apr 1;176 (7) :4419-30. PMID: 16547280). Recommended concentration: 1-3 ?g/mL.

CSMD1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CSMD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR