Anti-LPP Goat Polyclonal Antibody

Supplier: ProSci
42-271
10114-292EA 701.94 USD
10114-292
Anti-LPP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 85 kDa band observed in human placenta lysates and in HEPG2 lysate (calculated MW of 65.7 kDa according to NP_005569.1). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 0.1-0.3 ?g/mL.

LPP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR