Anti-FOXI3 Goat Polyclonal Antibody

Supplier: ProSci

42-264
10113-746EA 562.65 USD
10113-746
Anti-FOXI3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 50 kDa band in human skin lysates after 1 ?g/mL antibody staining . The 50 kDa band was successfully blocked by incubation with the immunizing peptide.

FOXI3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR