Anti-CCL20 Goat Polyclonal Antibody

Supplier: ProSci
42-213
10114-196EA 701.94 USD
10114-196
Anti-CCL20 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Immunohistochemistry: In paraffin embedded human liver shows local staining between the hepatocytes. Recommended concentration, 6-8 ?g/mL.

CCL20 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCL20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR