Anti-SUMF1 Goat Polyclonal Antibody

Supplier: ProSci

42-188
10114-160EA 560.36 USD
10114-160
Anti-SUMF1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 37-40 kDa band observed in mouse pancreas, mouse eye and human kidney lysates (calculated MW of 40.6 kDa according to human NP_877437.2 and 40.6 kDa according to mouse NP_666049.2). Recommended concentration: 0.5-2 ?g/mL.

SUMF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SUMF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR