Anti-PGF Goat Polyclonal Antibody

Supplier: ProSci
42-184
10114-154EA 689.28 USD
10114-154
Anti-PGF Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1000. Western Blot: Approximately 28 kDa band observed in human placenta lysates (calculated MW of 19.3 kDa according to NP_002623.2). The observed molecular weight corresponds to the glycosylated form. Recommended concentration: 1-3 ?g/mL.

PLGF antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR