Anti-PREX1 Goat Polyclonal Antibody

Supplier: ProSci

42-175
10114-142EA 560.21 USD
10114-142
Anti-PREX1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 150 kDa band observed in human brain (temporal cortex) lysates (calculated MW of 186 kDa according to NP_065871.2). Recommended concentration: 1-3 ?g/mL.

PREX1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PREX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR