Anti-MDFI Goat Polyclonal Antibody

Supplier: ProSci
42-165
10113-732EA 701.94 USD
10113-732
Anti-MDFI Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 70 kDa band in HEPG2 lysate after 0.3 ?g/mL antibody staining. The 70 kDa band was successfully blocked by incubation with the immunizing peptide.

MDFI antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MDFI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR