Anti-MTHFR Goat Polyclonal Antibody

Supplier: ProSci
42-160
10113-730EA 701.94 USD
10113-730
Anti-MTHFR Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 28 kDa band in human cerebellum, skeletal muscle and placenta lysates after 0.2 ?g/mL antibody staining. The 28 kDa band was successfully blocked by incubation with the immunizing peptide.

MTHFR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MTHFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR