Anti-HTR7 Goat Polyclonal Antibody

Supplier: ProSci
42-157
10114-124EA 701.94 USD
10114-124
Anti-HTR7 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 50 kDa band observed in lysates of cell line KELLY (calculated MW of 49.5 kDa according to NP_000863.1). Recommended concentration: 1-3 ?g/mL. an additional band of unknown identity was also consistently observed at 37 kDa . This band was successfully blocked by incubation with the immunizing peptide.

HTR7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HTR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR