Anti-UBE2F Goat Polyclonal Antibody

Supplier: ProSci
42-142
10114-096EA 701.94 USD
10114-096
Anti-UBE2F Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 20 kDa band observed in mouse liver lysates (calculated MW of 21.1 kDa according to human NP_542409.1 and to mouse NP_080730.1). Recommended concentration: 0.05-0.2 ?g/mL. Immunohistochemistry: In paraffin embedded human placenta shows membrane staining in trophoblasts-. Recommended concentration, 2.5-3.8 ?g/mL.

NCE2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: UBE2F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR