Anti-ASAM Goat Polyclonal Antibody

Supplier: ProSci
42-139
10114-092EA 701.94 USD
10114-092
Anti-ASAM Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 55 kDa band in human placenta lysates after 1 ?g/mL antibody staining.3 kDa according to NP_079045.1. The 55 kDa band was successfully blocked by incubation with the immunizing peptide.

CLMP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ASAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR