Anti-OTUD6A Goat Polyclonal Antibody

Supplier: ProSci
42-136
10114-090EA 552.17 USD
10114-090
Anti-OTUD6A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 22-23 kDa band in human pancreas, human ovary and human kidney lysates after 1 ?g/mL antibody staining. The 23 kDa band was successfully blocked by incubation with the immunizing peptide.

DUBA2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: OTUD6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR