Anti-HMGA1 Goat Polyclonal Antibody

Supplier: ProSci

42-115
10113-716EA 560.36 USD
10113-716
Anti-HMGA1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 24 kDa band observed in human pancreas (calculated MW of 11.7 kDa /10.7 kDa according to NP_665906.1; NP_002122.1). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. This band was successfully blocked by incubation with the immunizing peptide. Recommended concentration: 0.5-1 ?g/mL.

HMGA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HMGA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR