Anti-TMPRSS11D Goat Polyclonal Antibody

Supplier: ProSci
42-101
10114-044EA 552.17 USD
10114-044
Anti-TMPRSS11D Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 20 kDa band in lysates of HepG2 after 1 ?g/mL antibody staining. The 20 kDa band was successfully blocked by incubation with the immunizing peptide.

TMPRSS11D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TMPRSS11D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR