Anti-KLK6 Goat Polyclonal Antibody

Supplier: ProSci
42-098
10114-038EA 701.94 USD
10114-038
Anti-KLK6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 22 kDa band observed in mouse liver lysates (calculated MW of 27.5 kDa according to NP_035307.1). Recommended concentration: 1-3 ?g/mL. an additional band of unknown identity was also consistently observed at 48 kDa . Both bands were successfully blocked by incubation with the immunizing peptide.

Klk6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Klk6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR