Anti-SCGB3A1 Goat Polyclonal Antibody

Supplier: ProSci
42-085
10114-020EA 552.17 USD
10114-020
Anti-SCGB3A1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 33 kDa band in human breast tumor and normal lung lysates after 0.5 ?g/mL antibody staining.1 kDa according to NP_443095.2. The 33 kDa band was successfully blocked by incubation with the immunizing peptide.

SCGB3A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SCGB3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR