Anti-JAM2 Goat Polyclonal Antibody

Supplier: ProSci

42-074
10113-702EA 455.3 USD
10113-702
Anti-JAM2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 27 kDa band in human heart, liver and placenta lysates after 0.2 ?g/mL antibody staining. The 27 kDa band was successfully blocked by incubation with the immunizing peptide.

JAM2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: JAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR