Anti-RCBTB2 Goat Polyclonal Antibody

Supplier: ProSci
42-055
10113-968EA 701.94 USD
10113-968
Anti-RCBTB2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 30 kDa band in human liver, lung and tonsil lysates after 1 ?g/mL antibody staining.3 kDa according to NP_001259.1. The 30 kDa band was successfully blocked by incubation with the immunizing peptide.

RCBTB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RCBTB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR