Anti-AGXT Goat Polyclonal Antibody

Supplier: ProSci
42-054
10113-698EA 625.23 USD
10113-698
Anti-AGXT Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Preliminary experiments gave a faint 37 kDa band in human liver lysates after 1 ?g/mL antibody staining.0 kDa according to NP_000021.1. The 37 kDa band was successfully blocked by incubation with the immunizing peptide.

AGXT antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AGXT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR