Anti-C4ORF14 Goat Polyclonal Antibody

Supplier: ProSci

42-047
10113-956EA 455.3 USD
10113-956
Anti-C4ORF14 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 30 kDa band in human breast and in breast tumor lysates after 0.5 ?g/mL antibody staining.5 kDa according to NP_115689.1. The 30 kDa band was successfully blocked by incubation with the immunizing peptide.

C4orf14 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: C4orf14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR