Anti-APOL6 Goat Polyclonal Antibody

Supplier: ProSci

42-040
10113-692EA 560.36 USD
10113-692
Anti-APOL6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 38 kDa band observed in human kidney lysates (calculated MW of 38.1 kDa according to NP_085144.1). Recommended concentration: 1-3 ?g/mL.

APOL6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APOL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR