Anti-NUAK1 Goat Polyclonal Antibody

Supplier: ProSci
42-035
10113-936EA 701.94 USD
10113-936
Anti-NUAK1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 75 kDa band observed in human kidney, muscle and ovary lysates (calculated MW of 74.3 kDa according to NP_055655.1). Recommended concentration: 0.1-0.3 ?g/mL.

NUAK1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NUAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR