Anti-TREX2 Goat Polyclonal Antibody

Supplier: ProSci
42-027
10113-924EA 701.94 USD
10113-924
Anti-TREX2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 18 kDa band in lysates of Jurkat, MOLT4 and U937 after 0.5 ?g/mL antibody staining.9 kDa according to NP_542432.2. The 18 kDa band was successfully blocked by incubation with the immunizing peptide.

TREX2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TREX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR