Anti-Tankyrase 2 Goat Polyclonal Antibody

Supplier: ProSci
42-018
10113-912EA 701.94 USD
10113-912
Anti-Tankyrase 2 Goat Polyclonal Antibody
Antibodies
Western Blot: In transfected HEK293 transiently expressing TANK2 a band of approximately 127 kDa is observed. This band is not observed in the non-transfected HEK293. The calculated molecular size is 127 kDa according to NP_079511.1. Recommended concentration, 0.5-1.5 ?g/mL. Full image caption: HEK293 overexpressing TANK2 (lane 4) and TANK1 (lane 2) and probed with 42-018 (mock transfection in first lane). Lane three is empty. Lower panel shows the same lysates probed for alpha-Actin to show protein levels. Primary incubation (0.5 ?g/mL) was overnight at 4°C. Detected by chemiluminescence.

TNKS2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNKS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR