Anti-GPR81 Goat Polyclonal Antibody

Supplier: ProSci
42-015
10113-906EA 701.94 USD
10113-906
Anti-GPR81 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 40 kDa band observed in human liver lysates (calculated MW of 39.3 kDa according to NP_115943.1). Recommended concentration: 0.3-1 ?g/mL. an additional band of unknown identity was also consistently observed at 55 kDa . This band was successfully blocked by incubation with the immunizing peptide.

GPR81 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR81
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR