Anti-STK39 Goat Polyclonal Antibody

Supplier: ProSci
42-014
10113-904EA 701.94 USD
10113-904
Anti-STK39 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 65 kDa band observed in human cerebellum lysates and in Jurkat lysate (calculated MW of 59.5 kDa according to NP_037365.2). Recommended concentration: 0.3-1 ?g/mL.

STK39 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STK39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR