Anti-STK17B Goat Polyclonal Antibody

Supplier: ProSci
42-010
10113-896EA 701.94 USD
10113-896
Anti-STK17B Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 45 kDa band observed in Molt4 lysate (calculated MW of 42.3 kDa according to NP_004217.1). Recommended concentration: 1-3 ?g/mL.

DRAK2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STK17B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR