Anti-STIM2 Sheep Polyclonal Antibody

Supplier: ProSci
39-876
10070-720EA 647.67 USD
10070-720
Anti-STIM2 Sheep Polyclonal Antibody
STIM-2 is a component of the Ca sensor/channel. It associates with ORAI-1 and form a essential complex involved in store-operated channels. Expression of STIM and Orai proteins are found in many cell types and knock-out mice lack viability.

ELISA, Western (immuno) blotting, Immunostaining (in testing)

Affinity Purified

Type: Primary
Antigen: STIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR