Anti-PTRF Rabbit Polyclonal Antibody

Supplier: ProSci
39-860
10068-248EA 875.52 USD
10068-248
Anti-PTRF Rabbit Polyclonal Antibody
Antibodies
Cavin 2-SDR (serum deprivation response) is a member of the PKC enzyme family. Immunolocalization studies show Cavin 2 in caveoli. Cavin 2 appears to be recruited to caveoli by Cavin 1. Cavin 2 appears to be involved in some aspects of caveolar function yet to be determined

ELISA, Western (immuno) blotting, Immunostaining (in testing)

Affinity Purified

Type: Primary
Antigen: PTRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR