Anti-PTRF Rabbit Polyclonal Antibody

Supplier: ProSci
39-859
10068-246EA 875.52 USD
10068-246
Anti-PTRF Rabbit Polyclonal Antibody
Antibodies
Cavin 1-PTRF has been shown to be an essential component of caveolae. Cavin 1-PTRF appear to be recruited to membrane domains containing PS, cholesterol and oligomerized caveolins. Binding of Cavin 1-PTRF to these domains stabilizes the membrane curvature to produce the classic flask shape of caveolae. Disassembly of Cavin 1- PTRF releases caveolar components, including caveolins into the plasma membrane. Succinctly it appears that Cavin 1-PTRF regulates caveola structure and function.

ELISA, Western (immuno) blotting, Immunostaining (in testing)

Affinity Purified

Type: Primary
Antigen: PTRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR