Anti-BLM Rabbit Polyclonal Antibody

Supplier: ProSci

39-856
10068-242EA 700.46 USD
10068-242
Anti-BLM Rabbit Polyclonal Antibody
Antibodies
The Bloom syndrome gene product is a member of the RecQ helicase protein family. The protein contains 1417 amino acids; including one ATP binding site, one DEAH box, and two putative nuclear localization signals, as well as phosphorylation sites. BLS protein is known to associate and be phosphorylated by the ATR (ataxia telangiectasia, rad3+) protein, in response to genetic stress. The Bloom's syndrome gene product is thought to play a role in the etiology of two major human health problems, each of which is a very common complication of Bloom's syndrome, namely cancer and diabetes.

ELISA, Western (immuno) blotting, Immunostaining (in testing)

Affinity Purified

Type: Primary
Antigen: BLM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Non-Human Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR