Anti-TBC1D4 Rabbit Polyclonal Antibody

Supplier: ProSci
39-852
10068-236EA 875.52 USD
10068-236
Anti-TBC1D4 Rabbit Polyclonal Antibody
Antibodies
The Akt substrate 160 pT649, (TBC1D4-pT642) activation by Akt has been associated with the GLUT4 transloaction upon insulin activation in 3T3-L1 cells

ELISA, Western (immuno) blotting, Immunostaining (in testing)

Affinity Purified

Type: Primary
Antigen: TBC1D4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR