Anti-APP Rabbit Polyclonal Antibody

Supplier: ProSci
39-850
10068-232EA 875.52 USD
10068-232
Anti-APP Rabbit Polyclonal Antibody
Antibodies
Pyro-glutamic acid truncated amyloid beta 3-40/42 may have a greater propensity to aggregate under physiological conditions. In addition, this pyro E modification has been implicated as the molecular species responsible for seeding larger oligomers of amyloid-beta. Pyro-glutamic acid truncated amyloid beta 3-40/42’s exact involvement in the pathophysiology of Alzheimer’s disease has not been established.

  • ELISA , Western (immuno) blotting (in testing), Immunostaining (in testing)

Affinity Purified

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine, Sheep, Non-Human Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR