ELISA:<br>Sandwich:<br>In a sandwich ELISA (assuming 100?L/well), a concentration of 2.0-4.0 ?g/mL of this antibody will detect at least 500 pg/mL of recombinant Human Adiponectin when used in conjunction with compatible detection reagents at a concentration of approximately 0.5-1.0 ?g/mL. <br><br>Western Blot:<br><br>To detect Human Adiponectin by Western Blot analysis this antibody can be used at a concentration of 1.0-2.0 ?g/mL. When used in conjunction with compatible secondary reagents the detection limit for recombinant Human Adiponectin is 0.5-1.0 ng/lane, under reducing conditions and 1.0-2.0 ng/lane, under non-reducing conditions.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: 1.3-13A05-2D09
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human