Anti-SERPINI1 Rabbit Polyclonal Antibody (Biotin)

Supplier: ProSci
38-296
10098-038EA 576.13 USD
10098-038
Anti-SERPINI1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
ELISA:<br>Direct:<br>To detect Human Neuroserpin by direct ELISA (using 100 ml/well) a concentration of approximately 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody allows the detection of at least 0.2 – 0.4 ng/well of recombinant Human Neuroserpin. ELISA:<br>Sandwich:<br>To detect Human Neuroserpin by sandwich ELISA (using 100 ml/well) a concentration of 0.25 – 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with ProSci’s Polyclonal Anti- Human Neuroserpin (38-203) as a capture antibody, allows the detection of at least 0.2 – 0.4 ng/well of recombinant Human Neuroserpin. <br><br>Western Blot:<br><br>To detect Human Neuroserpin by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 mg/mL. When used in conjunction with compatible development reagents the detection limit for recombinant Human Neuroserpin is 1.5 – 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: SERPINI1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR