Anti-IL17B Rabbit Polyclonal Antibody

Supplier: ProSci

38-290
10098-028EA 455.16 USD
10098-028
Anti-IL17B Rabbit Polyclonal Antibody
Antibodies
ELISA:<br>Sandwich:<br>To detect hIL-17B by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with ProSci’s Polyclonal Biotinylated Anti-Human IL-17B (38-173) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hIL-17B. <br><br>Western Blot:<br><br>To detect hIL-17B by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hIL-17B is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: IL17B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR