Anti-IFNG Rabbit Polyclonal Antibody (Biotin)

Supplier: ProSci
38-257
10097-964EA 576.13 USD
10097-964
Anti-IFNG Rabbit Polyclonal Antibody (Biotin)
Antibodies
To detect Rat IFN-? by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant Rat IFN-? is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: Ifng
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR