Anti-NRG1 Rabbit Polyclonal Antibody

Supplier: ProSci
38-254
10097-958EA 576.13 USD
10097-958
Anti-NRG1 Rabbit Polyclonal Antibody
Antibodies
Neutralization:<br>To yield one-half maximal inhibition [ND50] of the biological activity of hHeregulin-B1 (1.0 ng/mL), a concentration of 0.1-1.1 ?g/mL of this antibody is required.<br><br>ELISA:<br><br>In a sandwich ELISA To detect hHeregulin-B1 (using 100 uL/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction withour biotinylated Anti-Human hHeregulin-B1as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hHeregulin-B1. <br><br>Western Blot:<br>To detect hHeregulin-B1 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ug/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hHeregulin-B1 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: NRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR