Anti-PTHLH Rabbit Polyclonal Antibody (Biotin)

Supplier: Prosci
38-209
10097-780EA 576.13 USD
10097-780
Anti-PTHLH Rabbit Polyclonal Antibody (Biotin)
Antibodies
ELISA:<br>Direct:<br>To detect hPTHrP by direct ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hPTHrP.<br><br>Sandwich<br>:To detect hPTHrP by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our polyclonal Anti-Human PTHrP as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hPTHrP. <br><br>Western Blot:<br>To detect hPTHrP by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hPTHrP is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: PTHLH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR