ELISA:<br>Direct:<br>To detect Rat VEGF by direct ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant Rat VEGF.<br><br>Sandwich<br>To detect Rat VEGF by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our polyclonal Anti-Rat VEGF as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant Rat VEGF. <br><br>Western Blot:<br>To detect Rat VEGF by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant Rat VEGF is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type: Primary
Antigen: Vegfa
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat